Merck
  • Evaluation of the antimicrobial, antioxidant, and anti-inflammatory activities of hydroxychavicol for its potential use as an oral care agent.
Evaluation of the antimicrobial, antioxidant, and anti-inflammatory activities of hydroxychavicol for its potential use as an oral care agent.
Antimicrobial agents and chemotherapy (2008-06-25)
Sandeep Sharma, Inshad Ali Khan, Intzar Ali, Furqan Ali, Manoj Kumar, Ashwani Kumar, Rakesh Kamal Johri, Sheikh Tasduq Abdullah, Sarang Bani, Anjali Pandey, Krishan Avtar Suri, Bishan Datt Gupta, Naresh Kumar Satti, Prabhu Dutt, Ghulam Nabi Qazi
ABSTRACT
Hydroxychavicol isolated from the chloroform extraction of aqueous extract of Piper betle leaves showed inhibitory activity against oral cavity pathogens. It exhibited an inhibitory effect on all of the oral cavity pathogens tested (MICs of 62.5 to 500 microg/ml) with a minimal bactericidal concentration that was twofold greater than the inhibitory concentration. Hydroxychavicol exhibited concentration-dependent killing of Streptococcus mutans ATCC 25175 up to 4x MIC and also prevented the formation of water-insoluble glucan. Interestingly, hydroxychavicol exhibited an extended postantibiotic effect of 6 to 7 h and prevented the emergence of mutants of S. mutans ATCC 25175 and Actinomyces viscosus ATCC 15987 at 2x MIC. Furthermore, it also inhibited the growth of biofilms generated by S. mutans and A. viscosus and reduced the preformed biofilms by these bacteria. Increased uptake of propidium iodide by hydroxychavicol-treated cells of S. mutans and A. viscosus indicated that hydroxychavicol probably works through the disruption of the permeability barrier of microbial membrane structures. Hydroxychavicol also exhibited potent antioxidant and anti-inflammatory activities. This was evident from its concentration-dependent inhibition of lipid peroxidation and significant suppression of tumor necrosis factor alpha expression in human neutrophils. Its efficacy against adherent cells of S. mutans in water-insoluble glucan in the presence of sucrose suggests that hydroxychavicol would be a useful compound for the development of antibacterial agents against oral pathogens and that it has great potential for use in mouthwash for preventing and treating oral infections.
Product Description
Supelco
Thymol, analytical standard
Sigma-Aldrich
Thymol, meets analytical specification of Ph. Eur., BP, NF, 99-101%
Sigma-Aldrich
Thymol, ≥98.5%
Sigma-Aldrich
Thymol, FCC, FG