Merck
  • Inhibition of Mitochondrial Calcium Overload by SIRT3 Prevents Obesity- or Age-Related Whitening of Brown Adipose Tissue.
Inhibition of Mitochondrial Calcium Overload by SIRT3 Prevents Obesity- or Age-Related Whitening of Brown Adipose Tissue.
Diabetes (2019-11-13)
Peng Gao, Yanli Jiang, Hao Wu, Fang Sun, Yaohong Li, Hongbo He, Bin Wang, Zongshi Lu, Yingru Hu, Xiao Wei, Yuanting Cui, Chengkang He, Lijuan Wang, Hongting Zheng, Gangyi Yang, Daoyan Liu, Zhencheng Yan, Zhiming Zhu
ABSTRACT
The whitening and loss of brown adipose tissue (BAT) during obesity and aging promote metabolic disorders and related diseases. The imbalance of Ca2+ homeostasis accounts for the dysfunction and clearance of mitochondria during BAT whitening. Capsaicin, a dietary factor activating TRPV1, can inhibit obesity induced by high-fat diet (HFD), but whether capsaicin inhibits BAT loss and the underlying mechanism remain unclear. In this study, we determined that the inhibitory effects of capsaicin on HFD-induced obesity and BAT whitening were dependent on the participation of SIRT3, a critical mitochondrial deacetylase. SIRT3 also mediated all of the beneficial effects of capsaicin on alleviating reactive oxygen species generation, elevating mitochondrial activity, and restricting mitochondrial calcium overload induced by HFD. Mechanistically, SIRT3 inhibits mitochondrial calcium uniporter (MCU)-mediated mitochondrial calcium overload by reducing the H3K27ac level on the MCU promoter in an AMPK-dependent manner. In addition, HFD also inhibits AMPK activity to reduce SIRT3 expression, which could be reversed by capsaicin. Capsaicin intervention also inhibited aging-induced BAT whitening through this mechanism. In conclusion, this study emphasizes a critical role of the AMPK/SIRT3 pathway in the maintenance of BAT morphology and function and suggests that intervention in this pathway may be an effective target for preventing obesity- or age-related metabolic diseases.
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Ru360, Ru360, is a cell-permeable oxygen-bridged dinuclear ruthenium amine complex. Binds to mitochondria with high affinity (Kd = 340 pM) and blocks Ca2+ uptake into mitochondria in vitro (IC₅₀ = 184 pM).