Merck
  • Cep128 associates with Odf2 to form the subdistal appendage of the centriole.
Cep128 associates with Odf2 to form the subdistal appendage of the centriole.
Genes to cells : devoted to molecular & cellular mechanisms (2019-01-10)
Hiroka Kashihara, Shuhei Chiba, Shin-Ichiro Kanno, Koya Suzuki, Tomoki Yano, Sachiko Tsukita
ABSTRACT
The mother centriole in a cell has two appendages, the distal appendage (DA) and subdistal appendage (SDA), which have roles in generating cilia and organizing the cellular microtubular network, respectively. In the knockout (KO) cells of Odf2, the component of the DA and SDA, both appendages simultaneously disappear. However, the molecular mechanisms by which the DA and SDA form independently but close to each other downstream of Odf2 are unknown. Here, using super-resolution structured illumination microscopy (SR-SIM), we found that the signal for GFP-tagged Odf2 overlapped considerably with that of immunofluorescently labeled Cep128. We further found that Cep128 knockdown (KD) caused the dissociation of other SDA components from the centriole, including centriolin, Ndel1, ninein and Cep170, whereas Odf2 was still associated with the centriole. In contrast, the DA components remained associated with the centriole in Cep128 KD cells. Consistent with this observation, we identified Cep128 as an Odf2-interacting protein by immunoprecipitation. Taken with the finding that Cep128 deletion decreased the stability of centriolar microtubules, our results indicate that Cep128 associates with Odf2 in the hierarchical assembly of SDA components to elicit the microtubule-organizing function.
Sigma-Aldrich
Anti-CNTRL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-CEP83 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-SCLT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-γ-Tubulin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-CEP89 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Ninein Antibody, clone 79-160-7, clone 79-160-7, from mouse