Merck
  • Apatinib inhibits gastric carcinoma development by regulating the expression levels of IL-17 via the Bax/Bcl-2 signaling pathway.
Apatinib inhibits gastric carcinoma development by regulating the expression levels of IL-17 via the Bax/Bcl-2 signaling pathway.
Experimental and therapeutic medicine (2021-05-11)
Tianxi Wang, Jun Zhang, Lihong Cui
ABSTRACT
Gastric carcinoma is a common type of gastrointestinal tumor with high morbidity and mortality rates. IL-17 is a newly discovered cytokine that has been reported to serve an important role in the development of gastric carcinoma. The potential effect of apatinib on IL-17 expression levels in the development of gastric carcinoma has been rarely reported. The present study aimed to investigate the potential mechanism of IL-17 and apatinib in the development of gastric carcinoma. A total of 30 tumor and para-carcinoma tissues were collected from 30 patients with gastric carcinoma between January 2019 and December 2019 and the expression levels of IL-17 in the tissues were analyzed by reverse transcription-quantitative PCR and western blotting. An in vitro model of gastric carcinoma was also established using the HGC-27 cell line, in which the cells were divided into control, IL-17, IL-17-apatinib and apatinib groups. The expression levels of IL-17, Bax, Bcl-2 and caspase-3 were analyzed using reverse transcription-quantitative PCR and western blotting. An MTT assay and flow cytometry were used to analyze the proliferation and apoptosis of HGC-27 cells, respectively, and a Transwell assay was used to analyze the invasive ability of HGC-27 cells. The results revealed that the expression levels of IL-17 were significantly upregulated in the gastric carcinoma tissues compared with the para-carcinoma tissues. In vitro, IL-17 treatment promoted the proliferation and invasive ability of HGC-27 cells, but inhibited the apoptosis with the significantly downregulated expression levels of Bax and caspase-3 and the upregulated expression levels of Bcl-2 than control group. Conversely, apatinib treatment significantly inhibited the proliferative and invasive abilities of HGC-27 cells, but promoted cell apoptosis in the IL-17 and IL-17-apatinib groups.. Collectively, the present results suggested that the upregulation of IL-17 may be associated with the occurrence and development of gastric carcinoma. The findings indicated that apatinib may inhibit gastric carcinoma development by regulating IL-17 expression via the Bax/Bcl-2 signaling pathway. Therefore, the present findings may enhance the current knowledge of the effect of apatinib on gastric carcinoma cells.
Sigma-Aldrich
Anti-IL-17 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-Bcl-2 antibody produced in mouse, clone Bcl-2-100, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Bax antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Pricing and availability is not currently available.