Merck
  • Non-isotopic dual parameter competition assay suitable for high-throughput screening of histone deacetylases.
Non-isotopic dual parameter competition assay suitable for high-throughput screening of histone deacetylases.
Bioorganic & medicinal chemistry letters (2009-05-22)
Daniel Riester, Christian Hildmann, Patricia Haus, Antonia Galetovic, Andreas Schober, Andreas Schwienhorst, Franz-Josef Meyer-Almes
ABSTRACT
Histone deacetylases reside among the most important and novel target classes in oncology. Selective lead structures are intensively developed to improve efficacy and reduce adverse effects. The common assays used so far to identify new lead structures suffer from many false positive hits due to auto-fluorescence of compounds or triggering undesired signal transduction pathways. These drawbacks are eliminated by the dual parameter competition assay reported in this study. The assay involves a new fluorescent inhibitor probe that shows an increase in both, fluorescence anisotropy and fluorescence lifetime upon binding to the enzyme. The assay is well suited for high-throughput screening.
Sigma-Aldrich
7-Amino-4-methylcoumarin, 99%
Sigma-Aldrich
7-Amino-4-methylcoumarin, Chromophore for substrates
Sigma-Aldrich
Atto 700, BioReagent, suitable for fluorescence, ≥90.0% (HPCE)
Sigma-Aldrich
Atto 700 NHS-ester, BioReagent, suitable for fluorescence, ≥90% (coupling to amine, HPLC)