Merck
  • Bis(dialkylaminethiocarbonyl)disulfides as potent and selective monoglyceride lipase inhibitors.
Bis(dialkylaminethiocarbonyl)disulfides as potent and selective monoglyceride lipase inhibitors.
Journal of medicinal chemistry (2009-11-04)
Coco N Kapanda, Giulio G Muccioli, Geoffray Labar, Jacques H Poupaert, Didier M Lambert
ABSTRACT
Monoglyceride lipase (MGL) inhibition may offer an approach in treating diseases in which higher 2-arachidonoyglycerol activity would be beneficial. We report here the synthesis and pharmacological evaluation of bis(dialkylaminethiocarbonyl)disulfide derivatives as irreversible MGL inhibitors. Inhibition occurs through interactions with MGL C208 and C242 residues, and these derivatives exhibit high inhibition selectivity over fatty acid amide hydrolase, another endocannabinoid-hydrolyzing enzyme.
Sigma-Aldrich
Tetramethylthiuram disulfide, 97%
Supelco
Thiram, PESTANAL®, analytical standard