Merck
  • Highly effective binding of viologens by p-sulfonatocalixarenes for the treatment of viologen poisoning.
Highly effective binding of viologens by p-sulfonatocalixarenes for the treatment of viologen poisoning.
Journal of medicinal chemistry (2009-10-16)
Kui Wang, Dong-Sheng Guo, Hong-Qing Zhang, Dong Li, Xi-Long Zheng, Yu Liu
ABSTRACT
Viologens are showing an increasing number of scientific and technical applications in addition to their use as herbicides. However, their high toxicity poses considerable risks to human health, society, and the environment. In this context, we propose a new therapeutic protocol for the treatment of viologen poisoning. The mechanism of this new protocol is based on host-guest chemistry and involves the effective inhibition of viologen toxicity by the complexation of p-sulfonatocalix[n]arenes. NMR, ITC, and X-ray crystallography studies indicated that p-sulfonatocalix[n]arenes could form highly stable complexes with viologens. Electrochemical results showed that the highly effective binding could induce the reduction potentials of viologens to shift to more negative values. Further studies in mice showed that the ingestion of p-sulfonatocalix[n]arenes significantly decreased the mortality rate of viologen-poisoned mice with lung and liver protection. As a result, p-sulfonatocalix[n]arenes may have potential application in the clinical treatment of viologen poisoning.
Sigma-Aldrich
Sodium salicylate, ReagentPlus®, ≥99.5% (titration)
Sigma-Aldrich
Sodium salicylate, 98.0-102.0% anhydrous basis, meets USP testing specifications
Sigma-Aldrich
Sodium salicylate, ≥99.5% (HPLC), puriss. p.a.
Sigma-Aldrich
Sodium salicylate, puriss. p.a., reag. Ph. Eur., 99.5-101.0% (calc. to the dried substance)