Merck
  • m6A Modification Prevents Formation of Endogenous Double-Stranded RNAs and Deleterious Innate Immune Responses during Hematopoietic Development.
m6A Modification Prevents Formation of Endogenous Double-Stranded RNAs and Deleterious Innate Immune Responses during Hematopoietic Development.
Immunity (2020-06-05)
Yimeng Gao, Radovan Vasic, Yuanbin Song, Rhea Teng, Chengyang Liu, Rana Gbyli, Giulia Biancon, Raman Nelakanti, Kirsten Lobben, Eriko Kudo, Wei Liu, Anastasia Ardasheva, Xiaoying Fu, Xiaman Wang, Poorval Joshi, Veronica Lee, Burak Dura, Gabriella Viero, Akiko Iwasaki, Rong Fan, Andrew Xiao, Richard A Flavell, Hua-Bing Li, Toma Tebaldi, Stephanie Halene
ABSTRACT
N6-methyladenosine (m6A) is the most abundant RNA modification, but little is known about its role in mammalian hematopoietic development. Here, we show that conditional deletion of the m6A writer METTL3 in murine fetal liver resulted in hematopoietic failure and perinatal lethality. Loss of METTL3 and m6A activated an aberrant innate immune response, mediated by the formation of endogenous double-stranded RNAs (dsRNAs). The aberrantly formed dsRNAs were long, highly m6A modified in their native state, characterized by low folding energies, and predominantly protein coding. We identified coinciding activation of pattern recognition receptor pathways normally tasked with the detection of foreign dsRNAs. Disruption of the aberrant immune response via abrogation of downstream Mavs or Rnasel signaling partially rescued the observed hematopoietic defects in METTL3-deficient cells in vitro and in vivo. Our results suggest that m6A modification protects against endogenous dsRNA formation and a deleterious innate immune response during mammalian hematopoietic development.
Product Description
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Anti-METTL14 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-phospho-PKR (pThr446) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-PKR antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Spermidine trihydrochloride, ≥98% (TLC)