Merck
  • Angiotensin II deteriorates advanced atherosclerosis by promoting MerTK cleavage and impairing efferocytosis through the AT1R/ROS/p38 MAPK/ADAM17 pathway.
Angiotensin II deteriorates advanced atherosclerosis by promoting MerTK cleavage and impairing efferocytosis through the AT1R/ROS/p38 MAPK/ADAM17 pathway.
American journal of physiology. Cell physiology (2019-08-08)
Yan Zhang, Ying Wang, Dong Zhou, Li-Sha Zhang, Fu-Xue Deng, Shan Shu, Li-Jun Wang, Yue Wu, Ning Guo, Juan Zhou, Zu-Yi Yuan
ABSTRACT
Vulnerable plaques in advanced atherosclerosis have defective efferocytosis. The role of ANG II in the progression of atherosclerosis is not fully understood. Herein, we investigated the effects and the underlying mechanisms of ANG II on macrophage efferocytosis in advanced atherosclerosis. ANG II decreased the surface expression of Mer tyrosine kinase (MerTK) in macrophages through a disintegrin and metalloproteinase17 (ADAM17)-mediated shedding of the soluble form of MerTK (sMer) in the medium, which led to efferocytosis suppression. ANG II-activated ADAM17 required reactive oxygen species (ROS) and p38 MAPK phosphorylation. Selective angiotensin II type 1 receptor (AT1R) blocker losartan suppressed ROS production, and ROS scavenger N-acetyl-l-cysteine (NAC) prevented p38 MAPK phosphorylation. In addition, mutant MERTKΔ483-488 was resistant to ANG II-induced MerTK shedding and efferocytosis suppression. The advanced atherosclerosis model that is characterized by larger necrotic cores, and less collagen content was established by feeding apolipoprotein E knockout (ApoE-/-) mice with a high-fat diet for 16 wk. NAC and losartan oral administration prevented atherosclerotic lesion progression. Meanwhile, the inefficient efferocytosis represented by decreased macrophage-associated apoptotic cells and decreased MerTK+CD68+double-positive macrophages in advanced atherosclerosis were prevented by losartan and NAC. Additionally, the serum levels of sMer were increased and positively correlated with the upregulated levels of ANG II in acute coronary syndrome (ACS) patients. In conclusion, ANG II promotes MerTK shedding via AT1R/ROS/p38 MAPK/ADAM17 pathway in macrophages, which led to defective efferocytosis and atherosclerosis progression. Defining the molecular mechanisms of defective efferocytosis may provide a promising prognosis and therapy for ACS patients.
Sigma-Aldrich
Fluorometric Intracellular Ros Kit, sufficient for 200 fluorometric tests (green)
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Sigma-Aldrich
Oil Red O, certified by the Biological Stain Commission
Sigma-Aldrich
SB 203580, solid, ≥98% (HPLC)
Sigma-Aldrich
InnoZyme TACE Activity Kit
Roche
In Situ Cell Death Detection Kit, TMR red, sufficient for ≤50 tests