Merck
  • Characterization of Biomarker Levels in Crimean-Congo Hemorrhagic Fever and Hantavirus Fever with Renal Syndrome.
Characterization of Biomarker Levels in Crimean-Congo Hemorrhagic Fever and Hantavirus Fever with Renal Syndrome.
Viruses (2019-07-31)
Miša Korva, Katarina Resman Rus, Miša Pavletič, Ana Saksida, Nataša Knap, Mateja Jelovšek, Katja Strašek Smrdel, Xhevat Jakupi, Isme Humolli, Jusuf Dedushaj, Miroslav Petrovec, Tatjana Avšič-Županc
ABSTRACT
Hemorrhagic fever with renal syndrome (HFRS) and Crimean-Congo hemorrhagic fever (CCHF) are important viral hemorrhagic fevers (VHF), especially in the Balkan region. Infections with Dobrava or Puumala orthohantavirus and Crimean-Congo hemorrhagic fever orthonairovirus can vary from a mild, nonspecific febrile illness, to a severe disease with a fatal outcome. The pathogenesis of both diseases is poorly understood, but it has been suggested that a host's immune mechanism might influence the pathogenesis of the diseases and survival. The aim of our study is to characterize cytokine response in patients with VHF in association with the disease progression and viral load. Forty soluble mediators of the immune response, coagulation, and endothelial dysfunction were measured in acute serum samples in 100 HFRS patients and 70 CCHF patients. HFRS and CCHF patients had significantly increased levels of IL-6, IL-12p70, IP-10, INF-γ, TNF-α, GM-CSF, MCP-3, and MIP-1b in comparison to the control group. Interestingly, HFRS patients had higher concentrations of serum MIP-1α, MIP-1β, which promote activation of macrophages and NK cells. HFRS patients had increased concentrations of IFN-γ and TNF-α, while CCHF patients had significantly higher concentrations of IFN-α and IL-8. In both, CCHF and HFRS patients' viral load significantly correlated with IP-10. Patients with fatal outcome had significantly elevated concentrations of IL-6, IFN-α2 and MIP-1α, while GRO-α, chemokine related to activation of neutrophils and basophils, was downregulated. Our study provided a comprehensive characterization of biomarkers released in the acute stages of CCHF and HFRS.
Product Description
Millipore
MILLIPLEX® Human Cardiovascular Disease (CVD) Magnetic Bead Panel 2 - Cardiovascular Disease Multiplex Assay, The analytes available for this multiplex kit are: ADAMTS13, D-Dimer, GDF-15, Myoglobin, sICAM-1, MPO, P-selectin, Lipocalin-2/NGAL, sVCAM-1, SAA.
Millipore
MILLIPLEX® Human Cardiovascular Disease (CVD) Magnetic Bead Panel 4 - Cardiovascular Disease Multiplex Assay, The analytes available for this multiplex kit are: sE-Selectin, Follistatin, Pecam-1, Pentraxin-3, Tissue Factor (TF), Thrombomodulin, Troponin T (TnT).
Millipore
MILLIPLEX® Human Cytokine/Chemokine Magnetic Bead Panel III - Immunology Multiplex Assay, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in mouse serum, plasma and cell culture samples.
Millipore
MILLIPLEX® Human Sepsis Magnetic Bead Panel 1 - Immune Response Multiplex Assay, Inflammation/Immunology Bead-Based Multiplex Assays using the Luminex technology enables the simultaneous analysis of multiple sepsis biomarkers in human serum, plasma and cell culture samples.
Millipore
MILLIPLEX® Human Cardiovascular Disease (Acute Phase) Magnetic Bead Panel 3 - Cardiovascular Disease Multiplex Assay, The analytes available for this multiplex kit are: Adipsin, AGP, α2-Macroglobulin, CRP, Fetuin A, Fibrinogen, L-Selectin, Serum Amyloid P, Haptoglobin, & Platelet Factor-4.
Millipore
MILLIPLEX® Human Angiogenesis/Growth Factor Magnetic Bead Panel - Cancer Multiplex Assay, Angiogenesie Bead-Based Multiplex Assays using the Luminex technology enables the simultaneous analysis of multiple angiogenic biomarkers in human serum, plasma and cell culture samples.