Merck
  • Reprogramming of host glutamine metabolism during Chlamydia trachomatis infection and its key role in peptidoglycan synthesis.
Reprogramming of host glutamine metabolism during Chlamydia trachomatis infection and its key role in peptidoglycan synthesis.
Nature microbiology (2020-08-05)
Karthika Rajeeve, Nadine Vollmuth, Sudha Janaki-Raman, Thomas F Wulff, Apoorva Baluapuri, Francesca R Dejure, Claudia Huber, Julian Fink, Maximilian Schmalhofer, Werner Schmitz, Rajeeve Sivadasan, Martin Eilers, Elmar Wolf, Wolfgang Eisenreich, Almut Schulze, Jürgen Seibel, Thomas Rudel
ABSTRACT
Obligate intracellular bacteria such as Chlamydia trachomatis undergo a complex developmental cycle between infectious, non-replicative elementary-body and non-infectious, replicative reticulate-body forms. Elementary bodies transform to reticulate bodies shortly after entering a host cell, a crucial process in infection, initiating chlamydial replication. As Chlamydia fail to replicate outside the host cell, it is unknown how the replicative part of the developmental cycle is initiated. Here we show, using a cell-free approach in axenic media, that the uptake of glutamine by the bacteria is crucial for peptidoglycan synthesis, which has a role in Chlamydia replication. The increased requirement for glutamine in infected cells is satisfied by reprogramming the glutamine metabolism in a c-Myc-dependent manner. Glutamine is effectively taken up by the glutamine transporter SLC1A5 and metabolized via glutaminase. Interference with this metabolic reprogramming limits the growth of Chlamydia. Intriguingly, Chlamydia failed to produce progeny in SLC1A5-knockout organoids and mice. Thus, we report on the central role of glutamine for the development of an obligate intracellular pathogenic bacterium and the reprogramming of host glutamine metabolism, which may provide a basis for innovative anti-infection strategies.
Sigma-Aldrich
10058-F4, ≥98% (HPLC), solid
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium, Without glucose, L-glutamine, phenol red, sodium pyruvate and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Meglumine diatrizoate, organic
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, sodium pyruvate, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Sodium diatrizoate hydrate, ≥98.0%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Slc1a7
Sigma-Aldrich
DL-2,6-Diaminopimelic acid, ≥95% (TLC)
Sigma-Aldrich
BPTES, ≥95% (HPLC)