Merck
  • Preclinical studies of ropivacaine extended-release from a temperature responsive hydrogel for prolonged relief of pain at the surgical wound.
Preclinical studies of ropivacaine extended-release from a temperature responsive hydrogel for prolonged relief of pain at the surgical wound.
International journal of pharmaceutics (2019-01-18)
Keun Sang Oh, Changsoon Hwang, Ha-Yeon Lee, Jin Sook Song, Hyun-Ji Park, Chul-Kyu Lee, InO Song, Tae-Hong Lim
ABSTRACT
Postoperative pain is a common form of acute pain that has been treated commonly by local anesthetics through regional nerve blocking. In this study, a series of experiments were conducted using rats to investigate the pharmacokinetic, distribution, and efficacy of a temperature responsive hydrogel-based drug delivery device (PF-72) containing ropivacaine (0.75%) for extended relief of postoperative pain by allowing the prolonged release of ropivacaine. When the ropivacaine was administered using PF-72, its concentration-time curve (AUClast) and peak concentration (Cmax) were 577.0 h*ng/mL and 271.9 ng/mL, respectively. In contrast when the ropivacaine solution was administered using saline solution, its AUClast and Cmax were 982.8 h*ng/mL and 423.6 ng/mL, respectively. In the tissue distribution study, the peak concentration and mean area under the curve of the ropivacaine in injection area (target tissue) were found about 2-fold higher in the case of PF-72 compared with the case of conventional ropivacaine solution. These results clearly demonstrate the capability of PF-72 hydrogel to retain the ropivacaine at the injection site for an extended period. Effective extended (at least 24 h) pain relief of ropivacaine administered using PF-72 was found in the pharmacodynamic study of prolonged analgesic effect. The results of this study indicated that local drug delivery by PF-72 hydrogel formulation may be an effective method to achieve extended relief of pain. Other advantages of ropivacaine administration using PF-72 include reduced systemic side effects and high localization of a drug in target tissues.