Merck
  • Complexation of the anti-Trypanosoma cruzi drug benznidazole improves solubility and efficacy.
Complexation of the anti-Trypanosoma cruzi drug benznidazole improves solubility and efficacy.
Journal of medicinal chemistry (2008-06-24)
Jean Jerley Nogueira Silva, Wander Rogério Pavanelli, Fredy R Salazar Gutierrez, Francisco Chagas Alves Lima, Albérico Borges Ferreira da Silva, João Santana Silva, Douglas Wagner Franco
ABSTRACT
The ruthenium complex, trans-[Ru(Bz)(NH 3) 4SO 2](CF 3SO 3) 2 1, Bz = benznidazole ( N-benzyl-2-(2-nitro-1 H-imidazol-1-yl)acetamide), is more hydrosoluble and more active (IC 50try/1 h = 79 +/- 3 microM) than free benznidazole 2 (IC 50try/1 h > 1 mM). 1 also exhibits low acute toxicity in vitro (IC 50macrophages > 1 mM) and in vivo (400 micromol/kg < LD 50 < 600 micromol/kg) and the formation of hydroxylamine is more favorable in 1 than in 2 by 9.6 kcal/mol. In murine acute models of Chagas' disease, 1 was more active than 2 even when only one dose was administrated. Moreover, 1 at a thousand-fold smaller concentration than the considered optimal dose for 2 (385 micromol/kg/day = 100 mg/kg/day), proved to be sufficient to protect all infected mice, eliminating the amastigotes in their hearts and skeletal muscles as observed in H&E micrographics.
Sigma-Aldrich
Gentian Violet, meets USP testing specifications
Sigma-Aldrich
Gentian violet, for microscopy (Bact., Hist.)
Sigma-Aldrich
Crystal Violet, indicator for the determination of the redox potential, S. No.: 785
Supelco
Crystal Violet, VETRANAL®, analytical standard
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
Crystal Violet Solution
Sigma-Aldrich
Crystal Violet Solution
Sigma-Aldrich
Crystal Violet, certified by the Biological Stain Commission
Sigma-Aldrich
Crystal Violet, for microscopy (Bact., Bot., Hist., Vit.), indicator (pH 0.1-2.0)