Merck
  • hESC-Derived Thalamic Organoids Form Reciprocal Projections When Fused with Cortical Organoids.
hESC-Derived Thalamic Organoids Form Reciprocal Projections When Fused with Cortical Organoids.
Cell stem cell (2019-02-26)
Yangfei Xiang, Yoshiaki Tanaka, Bilal Cakir, Benjamin Patterson, Kun-Yong Kim, Pingnan Sun, Young-Jin Kang, Mei Zhong, Xinran Liu, Prabir Patra, Sang-Hun Lee, Sherman M Weissman, In-Hyun Park
ABSTRACT
Human brain organoid techniques have rapidly advanced to facilitate investigating human brain development and diseases. These efforts have largely focused on generating telencephalon due to its direct relevance in a variety of forebrain disorders. Despite its importance as a relay hub between cortex and peripheral tissues, the investigation of three-dimensional (3D) organoid models for the human thalamus has not been explored. Here, we describe a method to differentiate human embryonic stem cells (hESCs) to thalamic organoids (hThOs) that specifically recapitulate the development of thalamus. Single-cell RNA sequencing revealed a formation of distinct thalamic lineages, which diverge from telencephalic fate. Importantly, we developed a 3D system to create the reciprocal projections between thalamus and cortex by fusing the two distinct region-specific organoids representing the developing thalamus or cortex. Our study provides a platform for understanding human thalamic development and modeling circuit organizations and related disorders in the brain.
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Anti-MAP2 Antibody, clone AP20, clone AP20, Chemicon®, from mouse
Sigma-Aldrich
Anti-Vesicular Glutamate Transporter 2 Antibody, clone 8G9.2, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-GFP IgG, CF 488A antibody produced in mouse, ~1 mg/mL, clone 9F9.F9, purified immunoglobulin