Merck
  • αV integrins in Schwann cells promote attachment to axons, but are dispensable in vivo.
αV integrins in Schwann cells promote attachment to axons, but are dispensable in vivo.
Glia (2020-08-04)
Kathleen K Catignas, Luciana R Frick, Marta Pellegatta, Edward Hurley, Zachary Kolb, Kathryn Addabbo, Joseph H McCarty, Richard O Hynes, Arjan van der Flier, Yannick Poitelon, Lawrence Wrabetz, Maria Laura Feltri
ABSTRACT
In the developing peripheral nervous system, Schwann cells (SCs) extend their processes to contact, sort, and myelinate axons. The mechanisms that contribute to the interaction between SCs and axons are just beginning to be elucidated. Using a SC-neuron coculture system, we demonstrate that Arg-Gly-Asp (RGD) peptides that inhibit αV -containing integrins delay the extension of SCs elongating on axons. αV integrins in SC localize to sites of contact with axons and are expressed early in development during radial sorting and myelination. Short interfering RNA-mediated knockdown of the αV integrin subunit also delays SC extension along axons in vitro, suggesting that αV -containing integrins participate in axo-glial interactions. However, mice lacking the αV subunit in SCs, alone or in combination with the potentially compensating α5 subunit, or the αV partners β3 or β8 , myelinate normally during development and remyelinate normally after nerve crush, indicating that overlapping or compensatory mechanisms may hide the in vivo role of RGD-binding integrins.
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
1,2-Dilinoleoyl-3-palmitoyl-rac-glycerol, ≥95% (TLC), liquid
Sigma-Aldrich
Anti-Integrin α5 Antibody, CT, Intracellular, serum, Chemicon®
TSKgel® ODS-100V HPLC Column, phase C18 (octadecyl), L × I.D. 25 cm × 2 mm, 3 μm particle size
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit
Sigma-Aldrich
Complement, Rabbit Serum, Complement, Rabbit Serum, is tested for suitability for HLA-ABC serology. Useful as an active source of complement for use with hybridoma antibodies, alloantisera, and related applications.
Sigma-Aldrich
Monoclonal Anti-Sodium Channel, Pan antibody produced in mouse, ~1 mg/mL, clone K58/35, purified immunoglobulin
Sigma-Aldrich
Anti-Calnexin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid
Sigma-Aldrich
Anti-Integrin alphaV Antibody, CT, Intracellular, serum, Chemicon®
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Fibronectin human plasma, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Toluidine Blue, 8.74% (ZN (THEORY)), for microscopy (Hist., Vit.)
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture