Merck
  • JAGN1 is required for fungal killing in neutrophil extracellular traps: Implications for severe congenital neutropenia.
JAGN1 is required for fungal killing in neutrophil extracellular traps: Implications for severe congenital neutropenia.
Journal of leukocyte biology (2018-08-15)
Avinash Khandagale, Beatrice Lazzaretto, Göran Carlsson, Mikael Sundin, Sulman Shafeeq, Ute Römling, Bengt Fadeel
ABSTRACT
Mutations in the gene JAGN1 were recently discovered in patients with severe congenital neutropenia (SCN). Neutrophils release neutrophil extracellular traps (NETs) consisting of decondensed chromatin decorated with various granular proteins such as neutrophil elastase and myeloperoxidase (MPO) to combat microbial infections. However, whether JAGN1 is required for the formation or function of NETs is not known. Here, we analyzed primary neutrophils from a patient with homozygous JAGN1 mutations with respect to phorbol myristate acetate (PMA)-induced NET formation. NET release was observed, but there appeared to be a reduced level of expression of MPO in the NETs. To study this further, we differentiated HL-60 cells into neutrophil-like cells and silenced JAGN1 expression by transfection with siRNA. These cells remained capable of producing NETs, but MPO expression was severely affected, and NETs released by JAGN1-silenced cells were ineffective in killing Candida albicans. The candidacidal function was restored upon treatment with GM-CSF or addition of MPO. GM-CSF also up-regulated the expression of calprotectin in NETs. Notably, JAGN1 did not impact on N-glycosylation of MPO in neutrophil-like HL-60 cells. These studies shed light on the susceptibility of SCN patients to fungal infections and the role of JAGN1 for the antimicrobial function of neutrophils exerted by NETs.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)−FITC antibody produced in sheep, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Neutrophil Elastase Rabbit pAb, liquid, Calbiochem®