Merck
  • Ca2+ Release by IP3 Receptors Is Required to Orient the Mitotic Spindle.
Ca2+ Release by IP3 Receptors Is Required to Orient the Mitotic Spindle.
Cell reports (2020-12-17)
Raul Lagos-Cabré, Adelina Ivanova, Colin W Taylor
ABSTRACT
The mitotic spindle distributes chromosomes evenly to daughter cells during mitosis. The orientation of the spindle, guided by internal and external cues, determines the axis of cell division and thereby contributes to tissue morphogenesis. Progression through mitosis requires local Ca2+ signals at critical steps, and because store-operated Ca2+ entry is inhibited during mitosis, those signals probably require Ca2+ release through inositol 1,4,5-trisphosphate receptors (IP3Rs). In cells without IP3Rs, astral microtubules around the daughter centrosome are shorter than those at the mother centrosome, and the mitotic spindle fails to align with the substratum during metaphase. The misalignment is due to the spindle ineffectively detecting internal cues rather than a failure of cells to recognize the substratum. Expression of type 3 IP3R is sufficient to rescue spindle alignment, but only if the IP3R has a functional pore. We conclude that Ca2+ signals evoked by IP3Rs are required to orient the mitotic spindle.
Sigma-Aldrich
MISSION® esiRNA, targeting human ITPR1
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial
Sigma-Aldrich
MISSION® esiRNA, targeting human ITPR3
Sigma-Aldrich
MISSION® esiRNA, targeting human ITPR2
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF405M antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution