Merck
  • Chemical synthesis and biological activities of 16alpha-derivatives of 5alpha-androstane-3alpha,17beta-diol as antiandrogens.
Chemical synthesis and biological activities of 16alpha-derivatives of 5alpha-androstane-3alpha,17beta-diol as antiandrogens.
Bioorganic & medicinal chemistry (2007-03-06)
Jenny Roy, Rock Breton, Céline Martel, Fernand Labrie, Donald Poirier
ABSTRACT
In our efforts to develop compounds with therapeutic potential as antiandrogens, we synthesized a series of 5alpha-androstane-3alpha,17beta-diol derivatives with a fixed side-chain length of 3-methylenes at C-16alpha, but bearing a diversity of functional groups at the end. Among these, the chloride induced the best antiproliferative activity on androgen-sensitive Shionogi cells. Substituting the OH at C-3 by a methoxy group showed the importance of the OH. Moreover, its transformation into a ketone increased the androgen receptor (AR) binding but decreased the antiproliferative activity and induced a proliferative effect on Shionogi cells. These results confirm the importance of keeping a 5alpha-androstane-3alpha,17beta-diol nucleus instead of a dihydrotestosterone nucleus. Variable side-chain lengths of 2-, 3-, 4-, and 6-methylenes at C-16alpha were investigated and the optimal length was found to be 3-methylenes. Although exhibiting a weak AR binding affinity, 16alpha-(3'-chloropropyl)-5alpha-androstane-3alpha,17beta-diol (15) provided an antiproliferative activity on Shionogi cells similar to that of pure non-steroidal antiandrogen hydroxy-flutamide (77% and 67%, respectively, at 0.1 microM). The new steroidal compound, 15, thus constitutes a good starting point for development of future antiandrogens with a therapeutic potential against prostate cancer.
Product Description
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Supelco
Testosterone solution, 1.0 mg/mL in 1,2-dimethoxyethane, drug standard