Merck
  • C4-Alkylamination of C4-Halo-1H-1-tritylpyrazoles Using Pd(dba)2 or CuI.
C4-Alkylamination of C4-Halo-1H-1-tritylpyrazoles Using Pd(dba)2 or CuI.
Molecules (Basel, Switzerland) (2020-10-16)
Yoshihide Usami, Yuya Tatsui, Hiroki Yoneyama, Shinya Harusawa
ABSTRACT
Alkylamino coupling reactions at the C4 positions of 4-halo-1H-1-tritylpyrazoles were investigated using palladium or copper catalysts. The Pd(dba)2 catalyzed C-N coupling reaction of aryl- or alkylamines, lacking a β-hydrogen atom, proceeded smoothly using tBuDavePhos as a ligand. As a substrate, 4-Bromo-1-tritylpyrazole was more effective than 4-iodo or chloro-1-tritylpyrazoles. Meanwhile, the CuI mediated C-N coupling reactions of 4-iodo-1H-1-tritylpyrazole were effective for alkylamines possessing a β-hydrogen atom.
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Sigma-Aldrich
1-Adamantylamine, 97%
Sigma-Aldrich
3-Phenyl-1-propylamine, 98%
Sigma-Aldrich
2-Acetylcyclohexanone, 97%
Sigma-Aldrich
2-Isobutyrylcyclohexanone, 96%