Merck
  • Evaluation of ultra-rapid susceptibility testing of ceftolozane-tazobactam by a flow cytometry assay directly from positive blood cultures.
Evaluation of ultra-rapid susceptibility testing of ceftolozane-tazobactam by a flow cytometry assay directly from positive blood cultures.
European journal of clinical microbiology & infectious diseases : official publication of the European Society of Clinical Microbiology (2020-06-03)
Inês Martins-Oliveira, Blanca Pérez-Viso, Sofia Quintas, Ana Silva-Dias, Rosário Gomes, Acácio G Rodrigues, Rafael Cantón, Cidalia Pina-Vaz
ABSTRACT
The urgent need for rapid antimicrobial susceptibility is broadly apparent from government reports to the lay press. Accordingly, we developed a flow-cytometry assay (FCM) for evaluating ceftolozane-tazobactam (C/T) susceptibility directly on blood cultures (BC) requiring < 2 h from flag positivity to report. The protocol was optimized with C/T-susceptible and C/T-resistant gram-negative bacilli inoculated in BC aerobic bottles (Becton-Dickinson, USA), and afterward optimized for different C/T concentrations (1/4, 2/4, 4/4, and 8/4 mg/L) for 1 h incubation (37 °C), followed by FCM and software analysis. Fluorescent membrane permeability and membrane potential dyes were comparatively used to detect early cell lesions using the CytoFLEX cytometer (Beckman-Coulter, USA). Repeatability, reproducibility, and stability of the assay up to 48 h after BC positivity were determined. Internal validation was performed in spiked BC bottles with 130 Enterobacterales and 32 Pseudomonas aeruginosa isolates from Porto University (Portugal), including 13 ATCC isolates. Additionally, 64 gram-negative bacilli recovered from positive BC at Ramon y Cajal Hospital (Madrid, Spain) were tested. Categorical agreement (CA) and analytical errors were calculated comparing FCM with broth microdilution results. Only the membrane potential dyes clearly distinguished CT-susceptible and CT-resistant isolates. Excellent repeatability, reproducibility, and inter-method concordance was observed. Overall, CA was 99.1% using EUCAST criteria with 2 major errors and 98.7% with CLSI criteria with 2 major and 1 minor errors. A new, accurate, and ultra-rapid FCM (< 2 h) for testing C/T susceptibility gave accurate results and would expand current FCM antimicrobial susceptibility assay.
Product Description
Millipore
Mueller Hinton Broth 2, NutriSelect® Plus, Cation-adjusted, suitable for microbiology
Sigma-Aldrich
Bis(1,3-dibutylbarbituric acid) trimethine oxonol, ≥95% (HPLC)