Merck
  • Decellularized extracellular matrix scaffolds identify full-length collagen VI as a driver of breast cancer cell invasion in obesity and metastasis.
Decellularized extracellular matrix scaffolds identify full-length collagen VI as a driver of breast cancer cell invasion in obesity and metastasis.
Science advances (2020-10-23)
Andrew L Wishart, Sydney J Conner, Justinne R Guarin, Jackson P Fatherree, Yifan Peng, Rachel A McGinn, Rebecca Crews, Stephen P Naber, Martin Hunter, Andrew S Greenberg, Madeleine J Oudin
ABSTRACT
The extracellular matrix (ECM), a major component of the tumor microenvironment, promotes local invasion to drive metastasis. Here, we describe a method to study whole-tissue ECM effects from disease states associated with metastasis on tumor cell phenotypes and identify the individual ECM proteins and signaling pathways that are driving these effects. We show that decellularized ECM from tumor-bearing and obese mammary glands drives TNBC cell invasion. Proteomics of the ECM from the obese mammary gland led us to identify full-length collagen VI as a novel driver of TNBC cell invasion whose abundance in tumor stroma increases with body mass index in human TNBC patients. Last, we describe the mechanism by which collagen VI contributes to TNBC cell invasion via NG2-EGFR cross-talk and MAPK signaling. Overall, these studies demonstrate the value of decellularized ECM scaffolds obtained from tissues to identify novel functions of the ECM.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Hematoxylin Solution, Gill No. 2
Sigma-Aldrich
Eosin Y Solution, Alcoholic
Sigma-Aldrich
Cholera Toxin from Vibrio cholerae, ≥90% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
Laminin from human placenta, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone P4C10, culture supernatant, clone P4C10, Chemicon®
Sigma-Aldrich
Tetraethylammonium borohydride, technical, ≥95% (T)
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid