Merck
NADPH oxidase 5 activation; a novel approach to human sperm cryoinjury.
Cell and tissue banking (2020-07-02)
Sara Keshtgar, Bahareh Ebrahimi, Seyed Mostafa Shid-Moosavi, Nasrollah Erfani
ABSTRACT
Sperm cryopreservation leads to various structural and functional damages, some of which induce by oxidative stress. The reactive oxygen species (ROS) generates by mitochondria and membrane NADPH oxidases (NOXs). Among the NOXs, only NOX5 has been identified in the cell membrane of human sperm. This study was designed to clarify the possible role of NOX5 on sperm cryoinjury. Forty human semen samples were washed and randomly divided into fresh and cryopreserved groups. Each group was divided into 4 subgroups containing Ham's F10 (control), 0.1% DMSO (vehicle), 100 nM of PMA (phorbol 12-myristate 13-acetate) and 1 µM of DPI (diphenyleneiodonium), as NOX5 activator and inhibitor. The samples of cryopreserved groups were preserved in liquid nitrogen for 1 month. The sperm kinematics, membrane integrity, ROS production, apoptosis rate, mitochondrial membrane potential (MMP), intracellular ATP and calcium concentration [Ca2+]i were evaluated. The percent of sperm with intact membrane and motile sperm reduced significantly after thawing (p ≤ 0.01). The ROS production (p ≤ 0.01) and the apoptotic rate increased, MMP dissipated, and the percentage of live cells with high [Ca2+]i decreased significantly in the cryopreserved control group relative to the fresh control group. DPI, in contrast to PMA, improved sperm progressive motility (p ≤ 0.01), membrane integrity in fresh and cryopreserved groups and reduced the ROS amount in cryopreserved group (p ≤ 0.01). Apoptotic rate, [Ca2+]i, ATP, and MMP did not change with DPI and PMA in cryopreserved groups. We conclude that NOX5 activity in fresh sperm is low, and it increases during cryopreservation. NOX5 inhibition improves the cryopreserved sperm quality.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Pluronic® F-127, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium citrate dihydrate, ≥99%, FG
Sigma-Aldrich
Peroxidase from horseradish, Type VI-A, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol), 950-2000 units/mg solid (using ABTS)
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Diphenyleneiodonium chloride, ≥98%
Sigma-Aldrich
Fluo 3-AM, ≥90% (TLC)