Merck
  • Lung-derived HMGB1 is detrimental for vascular remodeling of metabolically imbalanced arterial macrophages.
Lung-derived HMGB1 is detrimental for vascular remodeling of metabolically imbalanced arterial macrophages.
Nature communications (2020-08-29)
Ludovic Boytard, Tarik Hadi, Michele Silvestro, Hengdong Qu, Andrew Kumpfbeck, Rayan Sleiman, Kissinger Hyppolite Fils, Dornazsadat Alebrahim, Francesco Boccalatte, Matthias Kugler, Annanina Corsica, Bruce E Gelb, Glenn Jacobowitz, George Miller, Chiara Bellini, Jessica Oakes, Jean-Sébastien Silvestre, Lior Zangi, Bhama Ramkhelawon
ABSTRACT
Pulmonary disease increases the risk of developing abdominal aortic aneurysms (AAA). However, the mechanism underlying the pathological dialogue between the lungs and aorta is undefined. Here, we find that inflicting acute lung injury (ALI) to mice doubles their incidence of AAA and accelerates macrophage-driven proteolytic damage of the aortic wall. ALI-induced HMGB1 leaks and is captured by arterial macrophages thereby altering their mitochondrial metabolism through RIPK3. RIPK3 promotes mitochondrial fission leading to elevated oxidative stress via DRP1. This triggers MMP12 to lyse arterial matrix, thereby stimulating AAA. Administration of recombinant HMGB1 to WT, but not Ripk3-/- mice, recapitulates ALI-induced proteolytic collapse of arterial architecture. Deletion of RIPK3 in myeloid cells, DRP1 or MMP12 suppression in ALI-inflicted mice repress arterial stress and brake MMP12 release by transmural macrophages thereby maintaining a strengthened arterial framework refractory to AAA. Our results establish an inter-organ circuitry that alerts arterial macrophages to regulate vascular remodeling.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat epididymal adipocytes, Type II, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
HMG-1 human, lyophilized powder, ≥90% (SDS-PAGE), Histidine-tagged, recombinant, expressed in E. coli