Merck
  • Conserved Functions of Ether Lipids and Sphingolipids in the Early Secretory Pathway.
Conserved Functions of Ether Lipids and Sphingolipids in the Early Secretory Pathway.
Current biology : CB (2020-08-29)
Noemi Jiménez-Rojo, Manuel D Leonetti, Valeria Zoni, Adai Colom, Suihan Feng, Namrata R Iyengar, Stefan Matile, Aurélien Roux, Stefano Vanni, Jonathan S Weissman, Howard Riezman
ABSTRACT
Sphingolipids play important roles in physiology and cell biology, but a systematic examination of their functions is lacking. We performed a genome-wide CRISPRi screen in sphingolipid-depleted human cells and identified hypersensitive mutants in genes of membrane trafficking and lipid biosynthesis, including ether lipid synthesis. Systematic lipidomic analysis showed a coordinate regulation of ether lipids with sphingolipids, suggesting an adaptation and functional compensation. Biophysical experiments on model membranes show common properties of these structurally diverse lipids that also share a known function as glycosylphosphatidylinositol (GPI) anchors in different kingdoms of life. Molecular dynamics simulations show a selective enrichment of ether phosphatidylcholine around p24 proteins, which are receptors for the export of GPI-anchored proteins and have been shown to bind a specific sphingomyelin species. Our results support a model of convergent evolution of proteins and lipids, based on their physico-chemical properties, to regulate GPI-anchored protein transport and maintain homeostasis in the early secretory pathway.
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature, molecular biology grade
Sigma-Aldrich
Oleoyl chloride, ≥89%
Sigma-Aldrich
Myriocin from Mycelia sterilia, ≥98% (HPLC), powder
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
N,N-Diisopropylethylamine, purified by redistillation, 99.5%