Merck
  • Single-Cell RNA Sequencing of Human, Macaque, and Mouse Testes Uncovers Conserved and Divergent Features of Mammalian Spermatogenesis.
Single-Cell RNA Sequencing of Human, Macaque, and Mouse Testes Uncovers Conserved and Divergent Features of Mammalian Spermatogenesis.
Developmental cell (2020-06-07)
Adrienne Niederriter Shami, Xianing Zheng, Sarah K Munyoki, Qianyi Ma, Gabriel L Manske, Christopher D Green, Meena Sukhwani, Kyle E Orwig, Jun Z Li, Saher Sue Hammoud
ABSTRACT
Spermatogenesis is a highly regulated process that produces sperm to transmit genetic information to the next generation. Although extensively studied in mice, our current understanding of primate spermatogenesis is limited to populations defined by state-specific markers from rodent data. As between-species differences have been reported in the duration and differentiation hierarchy of this process, it remains unclear how molecular markers and cell states are conserved or have diverged from mice to man. To address this challenge, we employ single-cell RNA sequencing to identify transcriptional signatures of major germ and somatic cell types of the testes in human, macaque, and mice. This approach reveals similarities and differences in expression throughout spermatogenesis, including the stem/progenitor pool of spermatogonia, markers of differentiation, potential regulators of meiosis, RNA turnover during spermatid differentiation, and germ cell-soma communication. These datasets provide a rich foundation for future targeted mechanistic studies of primate germ cell development and in vitro gametogenesis.
Sigma-Aldrich
L-Arginine-13C6,15N4,2,3,3,4,4,5,5-d7 hydrochloride, 99 atom % 13C, 98 atom % 15N, 98 atom % D, 95% (CP)
Sigma-Aldrich
1H,1H,2H,2H-Perfluoro-1-octanol, 97%
Sigma-Aldrich
Anti-FMR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Ficoll® solution, Type 400, 20% in H2O
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
N-Lauroylsarcosine sodium salt solution, 20%, for molecular biology
Sigma-Aldrich
Anti-DNAJB6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-PIWIL4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Supelco
Busulfan, analytical standard, for drug analysis
Sigma-Aldrich
Anti-TCF3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Roche
DAB Substrate, dark brown, /black precipitate; Visually evaluated
Sigma-Aldrich
Anti-MAGEB2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody
Sigma-Aldrich
Triton X-100, for molecular biology
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg
Sigma-Aldrich
DAPI, for nucleic acid staining