Merck
  • Dioctatin Activates ClpP to Degrade Mitochondrial Components and Inhibits Aflatoxin Production.
Dioctatin Activates ClpP to Degrade Mitochondrial Components and Inhibits Aflatoxin Production.
Cell chemical biology (2020-09-06)
Tomohiro Furukawa, Hidekazu Katayama, Akira Oikawa, Lumi Negishi, Takuma Ichikawa, Michio Suzuki, Kohji Murase, Seiji Takayama, Shohei Sakuda
ABSTRACT
Aflatoxin contamination of crops is a serious problem worldwide. Utilization of aflatoxin production inhibitors is attractive, as the elucidation of their modes of action contributes to clarifying the mechanism of aflatoxin production. Here, we identified mitochondrial protease ClpP as the target of dioctatin, an inhibitor of aflatoxin production of Aspergillus flavus. Dioctatin conferred uncontrolled caseinolytic capacity on ClpP of A. flavus and Escherichia coli. Dioctatin-bound ClpP selectively degraded mitochondrial energy-related proteins in vitro, including a subunit of respiratory chain complex V, which was also reduced by dioctatin in a ClpP-dependent manner in vivo. Dioctatin enhanced glycolysis and alcohol fermentation while reducing tricarboxylic acid cycle metabolites. These disturbances were accompanied by reduced histone acetylation and reduced expression of aflatoxin biosynthetic genes. Our results suggest that dioctatin inhibits aflatoxin production by inducing ClpP-mediated degradation of mitochondrial energy-related components, and that mitochondrial energy metabolism functions as a key determinant of aflatoxin production.
Sigma-Aldrich
6-Nitroveratraldehyde, technical grade, 80%
Supelco
Aloe-emodin, analytical standard
Sigma-Aldrich
BIO, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
N-Succinyl-Leu-Tyr-7-amido-4-methylcoumarin, calpain substrate
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, sodium pyruvate, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
TPCA-1, ≥95% (HPLC)
Sigma-Aldrich
Ubiquitin from bovine erythrocytes, BioUltra, ≥98% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae, powder, ≥300 units/mg protein, mol wt ~141,000 (four subunits)
Sigma-Aldrich
Benzyl isocyanide, 98%
Millipore
Monoclonal Anti-HA−Agarose antibody produced in mouse, clone HA-7, purified immunoglobulin, PBS suspension
Sigma-Aldrich
4-Methoxyindole, 99%
Sigma-Aldrich
2-Nitrobenzyl alcohol, 97%