Merck
  • Generalized enzymatic mechanism of catalysis by tetrameric L-asparaginases from mesophilic bacteria.
Generalized enzymatic mechanism of catalysis by tetrameric L-asparaginases from mesophilic bacteria.
Scientific reports (2020-10-17)
Pawel Strzelczyk, Di Zhang, Marzena Dyba, Alexander Wlodawer, Jacek Lubkowski
ABSTRACT
The mechanism of catalysis by the L-glutaminase-asparaginase from Pseudomonas 7A (PGA) was investigated using structural, mass spectrometry, and kinetic data. We had previously proposed mechanism of hydrolysis of L-Asn by the type II L-asparaginase from E. coli (EcAII), but that work was limited to just one enzyme. Based on results presented in this report, we postulate that all homotetrameric L-asparaginases from mesophilic bacteria utilize a common ping-pong mechanism of catalysis consisting of two subsequent nucleophilic substitutions. Several new structures of non-covalent complexes of PGA with different substrates, as well as structures of covalent acyl-enzyme intermediates of PGA with canonical substrates (L-Asp and L-Glu) and an opportunistic ligand, a citrate anion, were determined. The results of kinetic experiments monitored by high-resolution LC/MS, when combined with new structural data, clearly show that the reaction catalyzed by L-glutaminase-asparaginases proceeds through formation of a covalent intermediate, as observed previously for EcAII. Additionally, by showing that the same mechanism applies to L-Asn and L-Gln, we postulate that it is common for all these structurally related enzymes.
Sigma-Aldrich
L-Aspartic acid β-hydroxamate, serine racemase inhibitor
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
8-Hydroxyquinoline
SAFC
Isopropyl β-D-1-thiogalactopyranoside