Merck
  • Development of Magnet-Driven and Image-Guided Degradable Microrobots for the Precise Delivery of Engineered Stem Cells for Cancer Therapy.
Development of Magnet-Driven and Image-Guided Degradable Microrobots for the Precise Delivery of Engineered Stem Cells for Cancer Therapy.
Small (Weinheim an der Bergstrasse, Germany) (2020-09-22)
Tanyong Wei, Jiang Liu, Dongfang Li, Shuxun Chen, Yachao Zhang, Junyang Li, Lei Fan, Zhangyan Guan, Chung-Mau Lo, Lidai Wang, Kwan Man, Dong Sun
ABSTRACT
Precise delivery of therapeutic cells to the desired site in vivo is an emerging and promising cellular therapy in precision medicine. This paper presents the development of a magnet-driven and image-guided degradable microrobot that can precisely deliver engineered stem cells for orthotopic liver tumor treatment. The microrobot employs a burr-like porous sphere structure and is made with a synthesized composite to fulfill degradability, mechanical strength, and magnetic actuation capability simultaneously. The cells can be spontaneously released from the microrobots on the basis of the optimized microrobot structure. The microrobot is actuated by a gradient magnetic field and guided by a unique photoacoustic imaging technology. In preclinical experiments on nude mice, microrobots carrying cells are injected via the portal vein and the released cells from the microrobots can inhibit the tumor growth greatly. This paper reveals for the first time of using degradable microrobots for precise delivery of therapeutic cells in vascular tissue and demonstrates its therapeutic effect in preclinical test.
Sigma-Aldrich
D-Luciferin potassium salt, ≥98.0% (HPLC)
Sigma-Aldrich
Pentaerythritol tetraacrylate, Pentaerythritol triacrylate, and Trimethylolpropane triacrylate mixture
Sigma-Aldrich
Poly(ethylene glycol) diacrylate, average Mn 575
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Toluene, anhydrous, 99.8%