Merck
Multicomponent DNA Polymerization Motor Gels.
Small (Weinheim an der Bergstrasse, Germany) (2020-08-11)
Ruohong Shi, Joshua Fern, Weinan Xu, Sisi Jia, Qi Huang, Gayatri Pahapale, Rebecca Schulman, David H Gracias
ABSTRACT
Hydrogels with the ability to change shape in response to biochemical stimuli are important for biosensing, smart medicine, drug delivery, and soft robotics. Here, a family of multicomponent DNA polymerization motor gels with different polymer backbones is created, including acrylamide-co-bis-acrylamide (Am-BIS), poly(ethylene glycol) diacrylate (PEGDA), and gelatin-methacryloyl (GelMA) that swell extensively in response to specific DNA sequences. A common mechanism, a polymerization motor that induces swelling is driven by a cascade of DNA hairpin insertions into hydrogel crosslinks. These multicomponent hydrogels can be photopatterned into distinct shapes, have a broad range of mechanical properties, including tunable shear moduli between 297 and 3888 Pa and enhanced biocompatibility. Human cells adhere to the GelMA-DNA gels and remain viable during ≈70% volumetric swelling of the gel scaffold induced by DNA sequences. The results demonstrate the generality of sequential DNA hairpin insertion as a mechanism for inducing shape change in multicomponent hydrogels, suggesting widespread applicability of polymerization motor gels in biomaterials science and engineering.
Sigma-Aldrich
Gelatin methacryloyl, gel strength 300 g Bloom, 80% degree of substitution
Sigma-Aldrich
Poly(ethylene glycol) diacrylate, average Mn 575
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
Sigma-Aldrich
Poly(ethylene glycol) diacrylate, average Mn 6,000, contains ≤1500 ppm MEHQ as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) diacrylate, average Mn 10,000, contains MEHQ as inhibitor
Sigma-Aldrich
TissueFab® Discrete GelMA, 300 bloom, 50% degree of substitution
Sigma-Aldrich
TissueFab® Discrete GelMA, 170-195 bloom, 50% degree of substitution
Sigma-Aldrich
TissueFab® Discrete GelMA, 90-110 bloom, 50% degree of substitution
Sigma-Aldrich
TissueFab® Discrete GelMA, 300 bloom, 80% degree of substitution