Merck
  • Physico-chemical characterization of carvacrol loaded zein nanoparticles for enhanced anticancer activity and investigation of molecular interactions between them by molecular docking.
Physico-chemical characterization of carvacrol loaded zein nanoparticles for enhanced anticancer activity and investigation of molecular interactions between them by molecular docking.
International journal of pharmaceutics (2020-08-28)
Priyanka Shinde, Hina Agraval, Amit Kumar Srivastav, Umesh C S Yadav, Umesh Kumar
ABSTRACT
Carvacrol (CV), a monoterpene possesses wide range of biological activities but has limited application due to low aqueous solubility and poor bioavailability. To address this issue and enhance bioavailability and efficacy of carvacrol, lecithin stabilized zein nanoparticles were investigated. Precipitation method was used for synthesis of nanoparticles and characterized using various techniques. CV entrapped under optimized parameters has size around 250 nm with -15 mV zeta potential. SEM studies showed nanoparticles with spherical morphology and size in accordance with DLS studies. FTIR, NMR and DSC were used to determine the molecular interaction between CV and lecithin stabilized zein nanoparticles. Molecular docking studies were performed to understand the interaction between protein and drug at molecular level. Our results demonstrated the presence of two active sites within zein, showing strong binding interactions with carvacrol. The encapsulation efficiency of 78% with loading efficiency of 13% was obtained as per HPLC and UV-Vis studies. Cytotoxicity assay indicated that the CV loaded nanoparticles induce cytotoxicity against colon cancer (SW480) cells further confirmed by acridine orange and ethidium bromide dual staining assay. Fluorescent tagged nanoparticles revealed significant cellular uptake of drug. Our results suggest that CV can be conveniently delivered via oral route after incorporating into lecithin stabilized zein nanoparticles and may prove effective for colon cancer treatment.
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Milli-Q
Milli-Q® IQ 7010 Ultrapure and Pure Water Purification System, Produces ultrapure (Type 1) water and pure (Type 2) water with a production flow rate of 10 L/hr from tap water feed.
Milli-Q
Milli-Q® IQ 7003 Ultrapure and Pure Water Purification System, Produces ultrapure (Type 1) water and pure (Type 2) water with a production flow rate of 3 L/hr from tap water feed
Milli-Q
Milli-Q® IQ 7005 Ultrapure and Pure Water Purification System, Produces ultrapure (Type 1) water and pure (Type 2) water with a production flow rate of 5 L/hr from tap water feed.
Milli-Q
Milli-Q® IQ 7015 Ultrapure and Pure Water Purification System, Produces ultrapure (Type 1) water and pure (Type 2) water with a production flow rate of 15 L/hr from tap water feed.