Merck
  • In vitro expansion of single Lgr5+ liver stem cells induced by Wnt-driven regeneration.
In vitro expansion of single Lgr5+ liver stem cells induced by Wnt-driven regeneration.
Nature (2013-01-29)
Meritxell Huch, Craig Dorrell, Sylvia F Boj, Johan H van Es, Vivian S W Li, Marc van de Wetering, Toshiro Sato, Karien Hamer, Nobuo Sasaki, Milton J Finegold, Annelise Haft, Robert G Vries, Markus Grompe, Hans Clevers
ABSTRACT
The Wnt target gene Lgr5 (leucine-rich-repeat-containing G-protein-coupled receptor 5) marks actively dividing stem cells in Wnt-driven, self-renewing tissues such as small intestine and colon, stomach and hair follicles. A three-dimensional culture system allows long-term clonal expansion of single Lgr5(+) stem cells into transplantable organoids (budding cysts) that retain many characteristics of the original epithelial architecture. A crucial component of the culture medium is the Wnt agonist RSPO1, the recently discovered ligand of LGR5. Here we show that Lgr5-lacZ is not expressed in healthy adult liver, however, small Lgr5-LacZ(+) cells appear near bile ducts upon damage, coinciding with robust activation of Wnt signalling. As shown by mouse lineage tracing using a new Lgr5-IRES-creERT2 knock-in allele, damage-induced Lgr5(+) cells generate hepatocytes and bile ducts in vivo. Single Lgr5(+) cells from damaged mouse liver can be clonally expanded as organoids in Rspo1-based culture medium over several months. Such clonal organoids can be induced to differentiate in vitro and to generate functional hepatocytes upon transplantation into Fah(-/-) mice. These findings indicate that previous observations concerning Lgr5(+) stem cells in actively self-renewing tissues can also be extended to damage-induced stem cells in a tissue with a low rate of spontaneous proliferation.
Sigma-Aldrich
Periodic Acid-Schiff (PAS) Staining System
Sigma-Aldrich
Y-27632 dihydrochloride, ≥98% (HPLC)
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid
Sigma-Aldrich
Carbon tetrachloride, reagent grade, 99.9%
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Gastrin I human, ≥95% (HPLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture