Merck
  • SIRT6 cooperates with SIRT5 to regulate bovine preadipocyte differentiation and lipid metabolism via the AMPKα signaling pathway.
SIRT6 cooperates with SIRT5 to regulate bovine preadipocyte differentiation and lipid metabolism via the AMPKα signaling pathway.
Archives of biochemistry and biophysics (2020-01-12)
Jieyun Hong, Chugang Mei, Sayed Haidar Abbas Raza, Rajwali Khan, Gong Cheng, Linsen Zan
ABSTRACT
Preadipocyte differentiation and lipid synthesis are critical steps for intramuscular fat (IMF) deposition and lipid metabolism homeostasis. IMF content of beef not only determines the ratio of muscle to adipose, but also determines the beef quality, flavor, and sensory characteristics. Maintaining lipid metabolism homeostasis is the key means of preventing and treating diabetes, obesity, and other metabolic diseases. SIRT6, which is an ADP-ribosyltransferase and NAD+-dependent deacetylase of acetyl and long-chain fatty acyl groups, playing central roles in lipid and glucose metabolism, is closely related to the occurrence of diabetes and obesity caused by overnutrition and aging. This study was based on bovine preadipocyte differentiation and an obese mice model, and comprehensively used transcriptome sequencing (RNA-seq) and morphological identification methods to explore the effects of inhibition of SIRT6 on differentiation and lipid synthesis, and related molecular mechanisms. Additionally, the feedback synergistic regulation of SIRT5 and SIRT6 on differentiation and lipid deposition was analyzed. The results showed that in the differentiation process of bovine preadipocytes, inhibition of SIRT5 significantly promoted SIRT6 expression. In addition, SIRT6 inhibited bovine preadipocyte differentiation and lipid synthesis, cooperating with SIRT5 to decrease lipid deposition, and repressed cell cycle arrest of preadipocytes. Moreover, in vivo verification experiments also obtained consistent results. Furthermore, SIRT6 inhibited preadipocyte differentiation and lipid deposition by activating the adenosine monophosphate activated protein kinase alpha (AMPKα) pathway. The above results provided a novel approach for understanding the functions of SIRT6 in regulating bovine adipocyte differentiation and lipid metabolism, as well as a new target for the treatment of diabetes and obesity in a clinical setting.
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Dounce tissue grinder pestle, Small clearance, working volume 2 mL