Merck
  • A small molecule NRF2 activator BC-1901S ameliorates inflammation through DCAF1/NRF2 axis.
A small molecule NRF2 activator BC-1901S ameliorates inflammation through DCAF1/NRF2 axis.
Redox biology (2020-03-17)
Yanwen Chen, John W Evankovich, Travis B Lear, Ferhan Tuncer, Jason R Kennerdell, Daniel P Camarco, Morgan S Shishido, Yuan Liu, Bill B Chen
ABSTRACT
NRF2 is a master regulator of cellular anti-oxidant and anti-inflammatory responses, and strategies to augment NRF2-dependent responses may beneficial in many diseases. Basal NRF2 protein level is constrained by constitutive KEAP1-mediated degradation, but in the presence of electrophiles, NRF2 ubiquitination is inhibited. Impeded NRF2 degradation increases NRF2 protein, resulting in up-regulation of anti-oxidant gene transcription, and decreased inflammation. KEAP1-independent mechanisms regulating NRF2 stability have also been reported. Here we employed an HTS approach and identified a small molecule, BC-1901S, that stabilized NRF2 and increased its activity. BC-1901S activated NRF2 by inhibiting NRF2 ubiquitination in a KEAP1-independent manner. It further increased NRF2-dependent anti-oxidant gene transcription, and exhibited anti-inflammatory effects in vitro and in vivo. Further, we identified a new NRF2-interacting partner, DDB1 and CUL4 Associated Factor 1 (DCAF1), an E3 ligase that targeted NRF2 for proteasomal degradation. Mechanistically, BC-1901S directly bound to DCAF1 and disrupted NRF2/DCAF1 interaction, thus activating NRF2. These findings provide new insights in NRF2 biology and NRF2 based anti-inflammatory therapy.
Sigma-Aldrich
Dimethyl fumarate, 97%