Merck
  • Functional analyses of miRNA-146b-5p during myogenic proliferation and differentiation in chicken myoblasts.
Functional analyses of miRNA-146b-5p during myogenic proliferation and differentiation in chicken myoblasts.
BMC molecular and cell biology (2020-05-31)
Jeong Hyo Lee, Seo Woo Kim, Ji Seon Han, Seung Pyo Shin, Sang In Lee, Tae Sub Park
ABSTRACT
In the poultry and livestock industries, precise genetic information is crucial for improving economic traits. Thus, functional genomic studies help to generate faster, healthier, and more efficient animal production. Chicken myoblast cells, which are required for muscle development and regeneration, are particularly important because chicken growth is closely related to muscle mass. In this study, we induced expression of microRNA-146b-5p mediated by the piggyBac transposon system in primary chicken myoblast (pCM) cells. Subsequently, we analyzed and compared the proliferation and differentiation capacity and also examined the expression of related genes in regular pCM (rpCM) cells and pCM cells overexpressing miRNA-146b-5p (pCM-146b OE cells). pCM-146b OE cells showed increased proliferation and upregulated gene expression related to cell proliferation. In addition, next-generation sequencing analyses were performed to compare global gene expression patterns between rpCM cells and pCM-146b OE cells. We found that the higher proliferation in pCM-146b OE cells was the result of upregulation of gene sets related to the cell cycle. Moreover, miRNA-146b-5p overexpression had inhibitory effects on myotube differentiation in pCM cells. Collectively these results demonstrate that miR-146b-5p is closely related to the proliferation and differentiation of chicken myogenic cells as a modulator of post-transcription.