Merck
  • FBXW7 Triggers Degradation of KMT2D to Favor Growth of Diffuse Large B-cell Lymphoma Cells.
FBXW7 Triggers Degradation of KMT2D to Favor Growth of Diffuse Large B-cell Lymphoma Cells.
Cancer research (2020-05-01)
Rizwan Saffie, Nan Zhou, Delphine Rolland, Özlem Önder, Venkatesha Basrur, Sydney Campbell, Kathryn E Wellen, Kojo S J Elenitoba-Johnson, Brian C Capell, Luca Busino
ABSTRACT
Mature B-cell neoplasms are the fifth most common neoplasm. Due to significant heterogeneity at the clinical and genetic levels, current therapies for these cancers fail to provide long-term cures. The clinical success of proteasome inhibition for the treatment of multiple myeloma and B-cell lymphomas has made the ubiquitin pathway an important emerging therapeutic target. In this study, we assessed the role of the E3 ligase FBXW7 in mature B-cell neoplasms. FBXW7 targeted the frequently inactivated tumor suppressor KMT2D for protein degradation, subsequently regulating gene expression signatures related to oxidative phosphorylation (OxPhos). Loss of FBXW7 inhibited diffuse large B-cell lymphoma cell growth and further sensitized cells to OxPhos inhibition. These data elucidate a novel mechanism of regulation of KMT2D levels by the ubiquitin pathway and uncover a role of FBXW7 in regulating oxidative phosphorylation in B-cell malignancies. SIGNIFICANCE: These findings characterize FBXW7 as a prosurvival factor in B-cell lymphoma via degradation of the chromatin modifier KMT2D.
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
NAE Inhibitor, MLN4924
Sigma-Aldrich
Doxycycline hyclate
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution