Merck
  • Nanoclay-based 3D printed scaffolds promote vascular ingrowth ex vivo and generate bone mineral tissue in vitro and in vivo.
Nanoclay-based 3D printed scaffolds promote vascular ingrowth ex vivo and generate bone mineral tissue in vitro and in vivo.
Biofabrication (2020-04-08)
Gianluca Cidonio, Michael Glinka, Yang-Hee Kim, Janos M Kanczler, Stuart A Lanham, Tilman Ahlfeld, Anja Lode, Jonathan I Dawson, Michael Gelinsky, Richard O C Oreffo
ABSTRACT
Acellular soft hydrogels are not ideal for hard tissue engineering given their poor mechanical stability, however, in combination with cellular components offer significant promise for tissue regeneration. Indeed, nanocomposite bioinks provide an attractive platform to deliver human bone marrow stromal cells (HBMSCs) in three dimensions producing cell-laden constructs that aim to facilitate bone repair and functionality. Here we present the in vitro, ex vivo and in vivo investigation of bioprinted HBMSCs encapsulated in a nanoclay-based bioink to produce viable and functional three-dimensional constructs. HBMSC-laden constructs remained viable over 21 d in vitro and immediately functional when conditioned with osteogenic media. 3D scaffolds seeded with human umbilical vein endothelial cells (HUVECs) and loaded with vascular endothelial growth factor (VEGF) implanted ex vivo into a chick chorioallantoic membrane (CAM) model showed integration and vascularisation after 7 d of incubation. In a pre-clinical in vivo application of a nanoclay-based bioink to regenerate skeletal tissue, we demonstrated bone morphogenetic protein-2 (BMP-2) absorbed scaffolds produced extensive mineralisation after 4 weeks (p < 0.0001) compared to the drug-free and alginate controls. In addition, HBMSC-laden 3D printed scaffolds were found to significantly (p < 0.0001) support bone tissue formation in vivo compared to acellular and cast scaffolds. These studies illustrate the potential of nanoclay-based bioink, to produce viable and functional constructs for clinically relevant skeletal tissue regeneration.
Sigma-Aldrich
Naphthol AS-MX Phosphate Alkaline Solution, 0.25%
Sigma-Aldrich
Alginic acid sodium salt from brown algae, BioReagent, suitable for immobilization of micro-organisms
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP