Merck
  • MYC Regulation of D2HGDH and L2HGDH Influences the Epigenome and Epitranscriptome.
MYC Regulation of D2HGDH and L2HGDH Influences the Epigenome and Epitranscriptome.
Cell chemical biology (2020-02-27)
ZhiJun Qiu, An-Ping Lin, Shoulei Jiang, Sara M Elkashef, Jamie Myers, Subramanya Srikantan, Binu Sasi, John Z Cao, Lucy A Godley, Dinesh Rakheja, Yingli Lyu, Siyuan Zheng, Muniswamy Madesh, Yuzuru Shiio, Patricia L M Dahia, Ricardo C T Aguiar
ABSTRACT
Mitochondrial D2HGDH and L2HGDH catalyze the oxidation of D-2-HG and L-2-HG, respectively, into αKG. This contributes to cellular homeostasis in part by modulating the activity of αKG-dependent dioxygenases. Signals that control the expression/activity of D2HGDH/L2HGDH are presumed to broadly influence physiology and pathology. Using cell and mouse models, we discovered that MYC directly induces D2HGDH and L2HGDH transcription. Furthermore, in a manner suggestive of D2HGDH, L2HGDH, and αKG dependency, MYC activates TET enzymes and RNA demethylases, and promotes their nuclear localization. Consistent with these observations, in primary B cell lymphomas MYC expression positively correlated with enhancer hypomethylation and overexpression of lymphomagenic genes. Together, these data provide additional evidence for the role of mitochondria metabolism in influencing the epigenome and epitranscriptome, and imply that in specific contexts wild-type TET enzymes could demethylate and activate oncogenic enhancers.
Sigma-Aldrich
Dimethyl 2-oxoglutarate, 96%
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-TET1 antibody produced in mouse, clone GT1462, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
DMOG, ≥98% (HPLC)
Sigma-Aldrich
Anti-TET3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution