Merck
  • SK channel-mediated metabolic escape to glycolysis inhibits ferroptosis and supports stress resistance in C. elegans.
SK channel-mediated metabolic escape to glycolysis inhibits ferroptosis and supports stress resistance in C. elegans.
Cell death & disease (2020-04-25)
Inge E Krabbendam, Birgit Honrath, Benjamin Dilberger, Eligio F Iannetti, Robyn S Branicky, Tammo Meyer, Bernard Evers, Frank J Dekker, Werner J H Koopman, Julien Beyrath, Daniele Bano, Martina Schmidt, Barbara M Bakker, Siegfried Hekimi, Carsten Culmsee, Gunter P Eckert, Amalia M Dolga
ABSTRACT
Metabolic flexibility is an essential characteristic of eukaryotic cells in order to adapt to physiological and environmental changes. Especially in mammalian cells, the metabolic switch from mitochondrial respiration to aerobic glycolysis provides flexibility to sustain cellular energy in pathophysiological conditions. For example, attenuation of mitochondrial respiration and/or metabolic shifts to glycolysis result in a metabolic rewiring that provide beneficial effects in neurodegenerative processes. Ferroptosis, a non-apoptotic form of cell death triggered by an impaired redox balance is gaining attention in the field of neurodegeneration. We showed recently that activation of small-conductance calcium-activated K+ (SK) channels modulated mitochondrial respiration and protected neuronal cells from oxidative death. Here, we investigated whether SK channel activation with CyPPA induces a glycolytic shift thereby increasing resilience of neuronal cells against ferroptosis, induced by erastin in vitro and in the nematode C. elegans exposed to mitochondrial poisons in vivo. High-resolution respirometry and extracellular flux analysis revealed that CyPPA, a positive modulator of SK channels, slightly reduced mitochondrial complex I activity, while increasing glycolysis and lactate production. Concomitantly, CyPPA rescued the neuronal cells from ferroptosis, while scavenging mitochondrial ROS and inhibiting glycolysis reduced its protection. Furthermore, SK channel activation increased survival of C. elegans challenged with mitochondrial toxins. Our findings shed light on metabolic mechanisms promoted through SK channel activation through mitohormesis, which enhances neuronal resilience against ferroptosis in vitro and promotes longevity in vivo.
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Nystatin Suspension, suspension, 10,000 unit/mL in DPBS, BioReagent, suitable for cell culture
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile
Sigma-Aldrich
CyPPA, ≥98% (HPLC), solid
Sigma-Aldrich
Tetramethylrhodamine ethyl ester perchlorate, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%
Sigma-Aldrich
5-Fluoro-2′-deoxyuridine, thymidylate synthase inhibitor