Merck
  • Vascular permeability disruption explored in the proteomes of mouse lungs and human microvascular cells following acute bromine exposure.
Vascular permeability disruption explored in the proteomes of mouse lungs and human microvascular cells following acute bromine exposure.
American journal of physiology. Lung cellular and molecular physiology (2020-06-25)
Dylan R Addis, Saurabh Aggarwal, Stephen F Doran, Ming-Yuan Jian, Israr Ahmad, Kyoko Kojima, David A Ford, Sadis Matalon, James A Mobley
ABSTRACT
Bromine (Br2) is an organohalide found in nature and is integral to many manufacturing processes. Br2 is toxic to living organisms, and high concentrations can prove fatal. To meet industrial demand, large amounts of purified Br2 are produced, transported, and stored worldwide, providing a multitude of interfaces for potential human exposure through either accidents or terrorism. To identify the key mechanisms associated with acute Br2 exposure, we have surveyed the lung proteomes of C57BL/6 male mice and human lung-derived microvascular endothelial cells (HMECs) at 24 h following exposure to Br2 in concentrations likely to be encountered in the vicinity of industrial accidents. Global discovery proteomics applications combined with systems biology analysis identified robust and highly significant changes in proteins associated with three biological processes: 1) exosome secretion, 2) inflammation, and 3) vascular permeability. We focused on the latter, conducting physiological studies on isolated perfused lungs harvested from mice 24 h after Br2 exposure. These experiments revealed significant increases in the filtration coefficient (Kf) indicating increased permeability of the pulmonary vasculature. Similarly, confluent monolayers of Br2 and Br-lipid-treated HMECs exhibited differential levels of zona occludens-1 that were found to be dissociated from cell wall localization, an increase in phosphorylation and internalization of E-cadherin, as well as increased actin stress fiber formation, all of which are consistent with increased permeability. Taken as a whole, our discovery proteomics and systems analysis workflow, combined with physiological measurements of permeability, revealed both profound and novel biological changes that contribute to our current understanding of Br2 toxicity.
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
DL-Dithiothreitol, for molecular biology, ≥98% (HPLC), ≥99% (titration)
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Anti-phospho-ROCK2 (pTyr722) antibody produced in rabbit, affinity isolated antibody