Merck
  • A Selective Serotonin Reuptake Inhibitor, a Proton Pump Inhibitor, and Two Calcium Channel Blockers Inhibit Candida albicans Biofilms.
A Selective Serotonin Reuptake Inhibitor, a Proton Pump Inhibitor, and Two Calcium Channel Blockers Inhibit Candida albicans Biofilms.
Microorganisms (2020-05-24)
Clarissa J Nobile, Craig L Ennis, Nairi Hartooni, Alexander D Johnson, Matthew B Lohse
ABSTRACT
Biofilms formed by the human fungal pathogen Candida albicans are naturally resistant to many of the antifungal agents commonly used in the clinic. We screened a library containing 1600 clinically tested drug compounds to identify compounds that inhibit C. albicans biofilm formation. The compounds that emerged from the initial screen were validated in a secondary screen and then tested for (1) their abilities to disrupt mature biofilms and (2) for synergistic interactions with representatives of the three antifungal agents most commonly prescribed to treat Candida infections, fluconazole, amphotericin B, and caspofungin. Twenty compounds had antibiofilm activity in at least one of the secondary assays and several affected biofilms but, at the same concentration, had little or no effect on planktonic (suspension) growth of C. albicans. Two calcium channel blockers, a selective serotonin reuptake inhibitor, and an azole-based proton pump inhibitor were among the hits, suggesting that members of these three classes of drugs or their derivatives may be useful for treating C. albicans biofilm infections.
Sigma-Aldrich
5,7-Dichloro-8-quinolinol, 99%
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%