Merck
  • Robotically handled whole-tissue culture system for the screening of oral drug formulations.
Robotically handled whole-tissue culture system for the screening of oral drug formulations.
Nature biomedical engineering (2020-04-29)
Thomas von Erlach, Sarah Saxton, Yunhua Shi, Daniel Minahan, Daniel Reker, Farhad Javid, Young-Ah Lucy Lee, Carl Schoellhammer, Tina Esfandiary, Cody Cleveland, Lucas Booth, Jiaqi Lin, Hannah Levy, Sophie Blackburn, Alison Hayward, Robert Langer, Giovanni Traverso
ABSTRACT
Monolayers of cancer-derived cell lines are widely used in the modelling of the gastrointestinal (GI) absorption of drugs and in oral drug development. However, they do not generally predict drug absorption in vivo. Here, we report a robotically handled system that uses large porcine GI tissue explants that are functionally maintained for an extended period in culture for the high-throughput interrogation (several thousand samples per day) of whole segments of the GI tract. The automated culture system provided higher predictability of drug absorption in the human GI tract than a Caco-2 Transwell system (Spearman's correlation coefficients of 0.906 and 0.302, respectively). By using the culture system to analyse the intestinal absorption of 2,930 formulations of the peptide drug oxytocin, we discovered an absorption enhancer that resulted in a 11.3-fold increase in the oral bioavailability of oxytocin in pigs in the absence of cellular disruption of the intestinal tissue. The robotically handled whole-tissue culture system should help advance the development of oral drug formulations and might also be useful for drug screening applications.
Sigma-Aldrich
Teicoplanin
Millipore
Chlortetracycline Selective Supplement, suitable for microbiology
Sigma-Aldrich
Insulin human, recombinant, expressed in yeast (proprietary host)
Sigma-Aldrich
Thioflavin T, used as stain for amyloid
Tristearin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Carbetocin acetate, ≥95% (HPLC)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Sodium glycocholate hydrate, ≥95% (TLC)
Sigma-Aldrich
Oxytocin