Merck
Forward RNAi screens in human stem cells.
Methods in molecular biology (Clifton, N.J.) (2010-08-06)
Christine Karlsson, Jonas Larsson, Aurélie Baudet
ABSTRACT
Identifying the genes and pathways that regulate self-renewal and differentiation in somatic stem cells is a central goal in stem cell and cancer biology. Here, we describe a method for RNAi-based screens in primary human hematopoietic stem and progenitor cells. These cells are suitable targets for complex, selection-based screens using pooled lentiviral shRNA libraries. The screening approach is a promising new tool to dissect regulatory mechanisms in hematopoietic and somatic stem cells, in general, and may be particularly useful to identify gene targets and modifiers that can be further exploited in strategies for ex vivo stem cell expansion.
Sigma-Aldrich
MISSION® LentiPlex® Human Pooled shRNA Library TRC2, For Rapid, Convenient Genome-wide shRNA Screens
Sigma-Aldrich
MISSION® LentiPlex® Human Pooled shRNA Library TRC1.5, For Rapid, Convenient Genome-wide shRNA Screens
Sigma-Aldrich
MISSION® LentiPlex® Complete Human Pooled shRNA Library, For Rapid, Convenient Genome-wide shRNA Screens
Sigma-Aldrich
MISSION® LentiPlex® Mouse Pooled shRNA Library TRC2, For Rapid, Convenient Genome-wide shRNA Screens
Sigma-Aldrich
MISSION® LentiPlex® Mouse Pooled shRNA Library TRC1.5, For Rapid, Convenient Genome-wide shRNA Screens
Sigma-Aldrich
MISSION® LentiPlex® Human Pooled shRNA Library, For Rapid, Convenient Genome-wide shRNA Screens
Sigma-Aldrich
MISSION® LentiPlex® Complete Mouse Pooled shRNA Library, For Rapid, Convenient Genome-wide shRNA Screens