Merck
  • A small molecule G6PD inhibitor reveals immune dependence on pentose phosphate pathway.
A small molecule G6PD inhibitor reveals immune dependence on pentose phosphate pathway.
Nature chemical biology (2020-05-13)
Jonathan M Ghergurovich, Juan C García-Cañaveras, Joshua Wang, Emily Schmidt, Zhaoyue Zhang, Tara TeSlaa, Harshel Patel, Li Chen, Emily C Britt, Marta Piqueras-Nebot, Mari Carmen Gomez-Cabrera, Agustín Lahoz, Jing Fan, Ulf H Beier, Hahn Kim, Joshua D Rabinowitz
ABSTRACT
Glucose is catabolized by two fundamental pathways, glycolysis to make ATP and the oxidative pentose phosphate pathway to make reduced nicotinamide adenine dinucleotide phosphate (NADPH). The first step of the oxidative pentose phosphate pathway is catalyzed by the enzyme glucose-6-phosphate dehydrogenase (G6PD). Here we develop metabolite reporter and deuterium tracer assays to monitor cellular G6PD activity. Using these, we show that the most widely cited G6PD antagonist, dehydroepiandosterone, does not robustly inhibit G6PD in cells. We then identify a small molecule (G6PDi-1) that more effectively inhibits G6PD. Across a range of cultured cells, G6PDi-1 depletes NADPH most strongly in lymphocytes. In T cells but not macrophages, G6PDi-1 markedly decreases inflammatory cytokine production. In neutrophils, it suppresses respiratory burst. Thus, we provide a cell-active small molecule tool for oxidative pentose phosphate pathway inhibition, and use it to identify G6PD as a pharmacological target for modulating immune response.
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mg
Please contact Customer Service for Availability
€42.20