Merck
IL-1β dysregulates cGMP signaling in the newborn lung.
American journal of physiology. Lung cellular and molecular physiology (2020-05-07)
Ying Zhong, Kristina Bry, Jesse D Roberts
ABSTRACT
Cyclic guanosine monophosphate (cGMP) signaling is an important regulator of newborn lung function and development. Although cGMP signaling is decreased in many models of newborn lung injury, the mechanisms are poorly understood. We determined how IL-1β regulates the expression of the α1-subunit of soluble guanylate cyclase (sGCα1), a prime effector of pulmonary cGMP signaling. Physiologic levels of IL-1β were discovered to rapidly decrease sGCα1 mRNA expression in a human fetal lung fibroblast cell line (IMR-90 cells) and protein levels in primary mouse pup lung fibroblasts. This sGCα1 expression inhibition appeared to be at a transcriptional level; IL-1β treatment did not alter sGCα1 mRNA stability although it reduced sGCα1 promoter activity. TGFβ-activated kinase 1 (TAK1) was determined to be required for IL-1β's regulation of sGCα1 expression; TAK1 knockdown protected sGCα1 mRNA expression in IL-1β-treated IMR-90 cells. Moreover, heterologously expressed TAK1 was sufficient to decrease sGCα1 mRNA levels in those cells. Nuclear factor-kappaB (NF-κB) signaling played a critical role in the IL-1β-TAK1-sGCα1 regulatory pathway; chromatin immunoprecipitation studies demonstrated enhanced activated NF-kB subunit (RelA) binding to the sGCα1 promoter after IL-1β treatment unless were treated with an IκB kinase2 inhibitor. Also, this NF-kB signaling inhibition protected sGCα1 expression in IL-1β-treated fibroblasts. Lastly, using transgenic mice in which active IL-1β was conditionally expressed in lung epithelial cells, we established that IL-1β expression is sufficient to stimulate TAK1 and decrease sGCα1 protein expression in the newborn lung. Together these results detail the role and mechanisms by which IL-1β inhibits cGMP signaling in the newborn lung.
Sigma-Aldrich
MISSION® esiRNA, targeting EGFP
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Map3k7
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
PHA-408, ≥98% (HPLC)
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Anti-Guanylyl Cyclase α1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Anti-α-Tubulin Antibody, clone DM1A, clone DM1A, Upstate®, from mouse
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal ANTI-FLAG® BioM2 antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution