Merck
  • NAD+ Controls Circadian Reprogramming through PER2 Nuclear Translocation to Counter Aging.
NAD+ Controls Circadian Reprogramming through PER2 Nuclear Translocation to Counter Aging.
Molecular cell (2020-05-06)
Daniel C Levine, Heekyung Hong, Benjamin J Weidemann, Kathryn M Ramsey, Alison H Affinati, Mark S Schmidt, Jonathan Cedernaes, Chiaki Omura, Rosemary Braun, Choogon Lee, Charles Brenner, Clara Bien Peek, Joseph Bass
ABSTRACT
Disrupted sleep-wake and molecular circadian rhythms are a feature of aging associated with metabolic disease and reduced levels of NAD+, yet whether changes in nucleotide metabolism control circadian behavioral and genomic rhythms remains unknown. Here, we reveal that supplementation with the NAD+ precursor nicotinamide riboside (NR) markedly reprograms metabolic and stress-response pathways that decline with aging through inhibition of the clock repressor PER2. NR enhances BMAL1 chromatin binding genome-wide through PER2K680 deacetylation, which in turn primes PER2 phosphorylation within a domain that controls nuclear transport and stability and that is mutated in human advanced sleep phase syndrome. In old mice, dampened BMAL1 chromatin binding, transcriptional oscillations, mitochondrial respiration rhythms, and late evening activity are restored by NAD+ repletion to youthful levels with NR. These results reveal effects of NAD+ on metabolism and the circadian system with aging through the spatiotemporal control of the molecular clock.
Sigma-Aldrich
TERGITOL solution, Type NP-40, 70% in H2O
Sigma-Aldrich
β-Nicotinamide mononucleotide, ≥95% (HPLC)
Sigma-Aldrich
Di(N-succinimidyl) glutarate, ≥97.0% (CHN)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Perchloric acid, ACS reagent, 70%