Merck
  • "Para-retinal" Vector Administration into the Deep Vitreous Enhances Retinal Transgene Expression.
"Para-retinal" Vector Administration into the Deep Vitreous Enhances Retinal Transgene Expression.
Molecular therapy. Methods & clinical development (2020-07-23)
Yong Zeng, Ryan Boyd, Joshua Bartoe, Henry E Wiley, Dario Marangoni, Lisa L Wei, Paul A Sieving
ABSTRACT
Intravitreal administration for human adeno-associated vector (AAV) delivery is easier and less traumatic to ocular tissues than subretinal injection, but it gives limited retinal transduction. AAV vectors are large (about 4,000 kDa) compared with most intraocular drugs, such as ranibizumab (48 kDa), and the large size impedes diffusion to reach the retina from the usual injection site in the anterior/mid-vitreous. Intuitively, a preferred placement for the vector would be deep in the vitreous near the retina, which we term "para-retinal" delivery. We explored the consequences of para-retinal intravitreal delivery in the rabbit eye and in non-human primate (NHP) eye. 1 h after para-retinal administration in the rabbit eye, the vector concentration near the retina remained four times greater than in the anterior vitreous, indicating limited vector diffusion through the gelatinous vitreous matrix. In NHP, para-retinal placement showed greater transduction in the fovea than vector applied in the mid-vitreous. More efficient retinal delivery translates to using lower vector doses, with reduced risk of ocular inflammatory exposure. These results indicate that para-retinal delivery yields more effective vector concentration near the retina, thereby increasing the potential for better retinal transduction in human clinical application.
Sigma-Aldrich
Anti-Granzyme B Antibody, clone GrB-7, culture supernatant, clone GrB-7, Chemicon®
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid