Merck
Small-Molecule PROTACS: New Approaches to Protein Degradation.
Angewandte Chemie (International ed. in English) (2016-01-13)
Momar Toure, Craig M Crews
ABSTRACT
The current inhibitor-based approach to therapeutics has inherent limitations owing to its occupancy-based model: 1) there is a need to maintain high systemic exposure to ensure sufficient in vivo inhibition, 2) high in vivo concentrations bring potential for off-target side effects, and 3) there is a need to bind to an active site, thus limiting the drug target space. As an alternative, induced protein degradation lacks these limitations. Based on an event-driven model, this approach offers a novel catalytic mechanism to irreversibly inhibit protein function by targeting protein destruction through recruitment to the cellular quality control machinery. Prior protein degrading strategies have lacked therapeutic potential. However, recent reports of small-molecule-based proteolysis-targeting chimeras (PROTACs) have demonstrated that this technology can effectively decrease the cellular levels of several protein classes.
Sigma-Aldrich
(S,R,S)-AHPC-CO-PEG4-C2-acid, ≥95%
Sigma-Aldrich
(S,R,S)-AHPC-acetamido-O-PEG4-C1-acid, ≥95%
Sigma-Aldrich
Pomalidomide-piperazine, ≥ 95.0%
Sigma-Aldrich
(S,R,S)-AHPC-CO-PEG1-C2-acid, ≥95%
Sigma-Aldrich
Pomalidomide-C9-NH2 hydrochloride
Sigma-Aldrich
(S,R,S)-AHPC-PEG4-Alkyne
Sigma-Aldrich
Pomalidomide-PEG5-Alkyne, ≥95%
Sigma-Aldrich
(S,R,S)-AHPC-PEG6-Azide, ≥95%
Sigma-Aldrich
Pomalidomide-PEG4-Azide, ≥95%
Sigma-Aldrich
Pomalidomide-C3-CO2H, ≥95%
Sigma-Aldrich
(S,R,S)-AHPC-PEG1-Alkyne, ≥95%
Sigma-Aldrich
Pomalidomide-PEG5-NH2 hydrochloride
Sigma-Aldrich
2,7-Diazaspiro[3.5]nonane-7-acetic acid, 2-[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-5-yl]-, ≥95%
Sigma-Aldrich
Thalidomide-O-amido-C8-NH2 trifluoroacetate, ≥95.0%
Sigma-Aldrich
Pomalidomide-PEG2-C2-azide, ≥95%
Sigma-Aldrich
VH 032 amide-alkyl C5-acid, ≥95%
Sigma-Aldrich
(S,R,S)-AHPC-C6-PEG3-butyl amine hydrochloride, ≥95%
Sigma-Aldrich
4-Pentynoic acid, 5-[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1-oxo-1H-isoindol-4-yl], ≥95.0%
Sigma-Aldrich
(S,R,S)-AHPC-PEG6-NH2 hydrochloride, ≥95%
Sigma-Aldrich
(S,R,S)-AHPC-PEG5-Alkyne
Sigma-Aldrich
Pomalidomide-PEG4-NH2 hydrochloride, ≥95%
Sigma-Aldrich
3-Pyridinecarboxylic acid, 6-[4-[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-5-yl]-1-piperazinyl]-, ≥95%
Sigma-Aldrich
(S,R,S)-AHPC-piperazine-pyridine-alkyne-NH2 hydrochloride
Sigma-Aldrich
Propanoic acid, 3-[2-[2-[2-[[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-5-yl]amino]ethoxy]ethoxy]ethoxy]-, ≥95%
Sigma-Aldrich
Pomalidomide-C2-NH2 hydrochloride, ≥95%
Sigma-Aldrich
(S,R,S)-AHPC-pentanoic-acid, ≥95%
Sigma-Aldrich
VH032-cyclopropane-F, ≥95%
Sigma-Aldrich
FBnG-C3-PEG5-C3-NH2 hydrochloride, ≥95%
Sigma-Aldrich
(S,R,S)-AHPC-C9-NH2 hydrochloride, ≥95%
Sigma-Aldrich
Pomalidomide-difluoroPEG1-C4-piperazine Hydrochloride, ≥95%