Merck
  • Black Lipid Membranes: Challenges in Simultaneous Quantitative Characterization by Electrophysiology and Fluorescence Microscopy.
Black Lipid Membranes: Challenges in Simultaneous Quantitative Characterization by Electrophysiology and Fluorescence Microscopy.
Langmuir : the ACS journal of surfaces and colloids (2019-06-28)
Maria Tsemperouli, Esther Amstad, Naomi Sakai, Stefan Matile, Kaori Sugihara
ABSTRACT
Horizontal black lipid membranes (BLMs) enable optical microscopy to be combined with the electrophysiological measurements for studying ion channels, peptide pores, and ionophores. However, a careful literature review reveals that simultaneous fluorescence and electrical recordings in horizontal BLMs have been rarely reported for an unclear reason, whereas many works employ bright-field microscopy instead of fluorescence microscopy or perform fluorescence imaging and electrical measurements one after another separately without truly exploiting the advantage of the combined setup. In this work, the major causes related to the simultaneous electrical and fluorescence recordings in horizontal BLMs are identified, and several solutions to counteract the issue are also proposed.
Product Description
Avanti
18:1 Biotinyl Cap PE, Avanti Researchā„¢ - A Croda Brand 870273P, powder
Avanti
18:1 Biotinyl Cap PE, Avanti Researchā„¢ - A Croda Brand 870273C