Merck
  • Glutathione dynamics determine the therapeutic efficacy of mesenchymal stem cells for graft-versus-host disease via CREB1-NRF2 pathway.
Glutathione dynamics determine the therapeutic efficacy of mesenchymal stem cells for graft-versus-host disease via CREB1-NRF2 pathway.
Science advances (2020-06-04)
Jisun Lim, Jinbeom Heo, Hyein Ju, Ji-Woong Shin, YongHwan Kim, Seungun Lee, Hwan Yeul Yu, Chae-Min Ryu, HongDuck Yun, Sujin Song, Ki-Sung Hong, Hyung-Min Chung, Hwa-Ryeon Kim, Jae-Seok Roe, Kihang Choi, In-Gyu Kim, Eui Man Jeong, Dong-Myung Shin
ABSTRACT
Glutathione (GSH), the most abundant nonprotein thiol functioning as an antioxidant, plays critical roles in maintaining the core functions of mesenchymal stem cells (MSCs), which are used as a cellular immunotherapy for graft-versus-host disease (GVHD). However, the role of GSH dynamics in MSCs remains elusive. Genome-wide gene expression profiling and high-throughput live-cell imaging assays revealed that CREB1 enforced the GSH-recovering capacity (GRC) of MSCs through NRF2 by directly up-regulating NRF2 target genes responsible for GSH synthesis and redox cycling. MSCs with enhanced GSH levels and GRC mediated by CREB1-NRF2 have improved self-renewal, migratory, anti-inflammatory, and T cell suppression capacities. Administration of MSCs overexpressing CREB1-NRF2 target genes alleviated GVHD in a humanized mouse model, resulting in improved survival, decreased weight loss, and reduced histopathologic damages in GVHD target organs. Collectively, these findings demonstrate the molecular and functional importance of the CREB1-NRF2 pathway in maintaining MSC GSH dynamics, determining therapeutic outcomes for GVHD treatment.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
AMD3100 octahydrochloride hydrate, ≥97% (NMR), solid
Sigma-Aldrich
L-Buthionine-sulfoximine, ≥97% (TLC)
Sigma-Aldrich
Glutathione reduced ethyl ester, ≥90% (TLC)
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human FOSL1
Sigma-Aldrich
Diamide
Sigma-Aldrich
Lectin from Phaseolus vulgaris (red kidney bean), Phytohemagglutinin PHA-M, lyophilized powder
Sigma-Aldrich
Crystal Violet, certified by the Biological Stain Commission