Merck
  • Abscisic acid rescues the root meristem defects of the Medicago truncatula latd mutant.
Abscisic acid rescues the root meristem defects of the Medicago truncatula latd mutant.
Developmental biology (2007-01-24)
Yan Liang, David M Mitchell, Jeanne M Harris
ABSTRACT
The LATD gene of the model legume, Medicago truncatula, is required for the normal function of three meristems, i.e. the primary root, lateral roots and nitrogen-fixing nodules. In latd mutants, primary root growth eventually arrests, resulting in a disorganized root tip lacking a presumptive meristem and root cap columella cells. Lateral root organs are more severely affected; latd lateral roots and nodules arrest immediately after emerging from the primary root, and reveal a lack of organization. Here we show that the plant hormone, abscisic acid (ABA), can rescue the latd root, but not nodule, meristem defects. Growth on ABA is sufficient to restore formation of small, cytoplasm-rich cells in the presumptive meristem region, rescue meristem organization and root growth and formation of root cap columella cells. In contrast, inhibition of ethylene synthesis or signaling fails to restore latd primary root growth. We find that latd mutants have normal levels of ABA, but exhibit reduced sensitivity to the hormone in two other ABA-dependent processes: seed germination and stomatal closure. Together, these observations demonstrate that the latd mutant is defective in the ABA response and indicate a role for LATD-dependent ABA signaling in M. truncatula root meristem function.
Sigma-Aldrich
(+)-Abscisic acid, ≥98% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Abscisic Acid Immunoassay Detection Kit, Quantitate the levels of abscisic acid (ABA) in plant tissue